ENA341295204 Enamine

2-chloro-N-(trimethylthiophen-2-yl)acetamide

  • MDL number MFCD20441763
  • Cas Number : 1354961-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-91679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91679
Availability details for
Quantity
  • Availability
  • Add to Cart