ENA341295209 Enamine

N-methyl-N-(2,2,2-trifluoroethyl)piperidine-3-carboxamide hydrochloride

  • MDL number MFCD20441766
  • Cas Number : 1354958-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91684

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91684
Availability details for
Quantity
  • Availability
  • Add to Cart