ENA341295215 Enamine

(2S)-1-phenylbutan-2-ol

  • MDL number MFCD20501960
  • Cas Number : 74709-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91692
Availability details for
Quantity
  • Availability
  • Add to Cart