ENA341295229 Enamine

2-[1-(6-chloropyridazin-3-yl)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl]acetic acid

  • MDL number MFCD11133564
  • Cas Number : 1094331-75-5
  • Molecular Weight : 254.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-91745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91745
Availability details for
Quantity
  • Availability
  • Add to Cart