ENA341295233 Enamine

1-(4,6-dimethylpyrimidin-2-yl)-3-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11180139
  • Cas Number : 1152508-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91750
Availability details for
Quantity
  • Availability
  • Add to Cart