ENA341295236 Enamine

N-{[3-(aminomethyl)phenyl]methyl}methanesulfonamide

  • MDL number MFCD12079064
  • Cas Number : 1153541-60-6
  • Molecular Weight : 214.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91765
Availability details for
Quantity
  • Availability
  • Add to Cart