ENA341295238 Enamine

1-(1-phenyl-1H-imidazole-5-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD13668333

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91768
Availability details for
Quantity
  • Availability
  • Add to Cart