ENA341295246 Enamine

2-(chloromethyl)-5-(2-methylphenyl)-1,3-oxazole

  • MDL number MFCD09835623
  • Cas Number : 1094226-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91778
Availability details for
Quantity
  • Availability
  • Add to Cart