ENA341295249 Enamine

ethyl 2-amino-2-methyl-3-phenylpropanoate

  • MDL number MFCD02663005
  • Cas Number : 46438-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91782
Availability details for
Quantity
  • Availability
  • Add to Cart