ENA341295250 Enamine

(2S)-2-methanesulfonamido-3-methylbutanoic acid

  • MDL number MFCD12795451
  • Cas Number : 97482-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91783
Availability details for
Quantity
  • Availability
  • Add to Cart