ENA341295253 Enamine

1-(4-methylphenyl)butan-2-ol

  • MDL number MFCD11934523
  • Cas Number : 110661-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91786
Availability details for
Quantity
  • Availability
  • Add to Cart