ENA341295254 Enamine

1-cyclopropyl-2-(4-methylphenyl)ethan-1-ol

  • MDL number MFCD11934749
  • Cas Number : 132713-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-91787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91787
Availability details for
Quantity
  • Availability
  • Add to Cart