ENA341295256 Enamine

2-(4-cyclobutanecarbonylpiperazin-1-yl)ethan-1-ol

  • MDL number MFCD12179904
  • Cas Number : 1155608-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91790

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91790
Availability details for
Quantity
  • Availability
  • Add to Cart