ENA341295266 Enamine

3-(4-amino-3,5-dimethyl-1H-pyrazol-1-yl)-1lambda6-thiolane-1,1-dione

  • MDL number MFCD11058207
  • Cas Number : 1156111-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-91828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91828
Availability details for
Quantity
  • Availability
  • Add to Cart