ENA341295280 Enamine

2-(chloromethyl)-5-(3-methoxyphenyl)-1,3,4-oxadiazole

  • MDL number MFCD08753714
  • Cas Number : 36770-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91851

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91851
Availability details for
Quantity
  • Availability
  • Add to Cart