ENA341295281 Enamine

N-(3-chlorophenyl)prop-2-enamide

  • MDL number MFCD00583235
  • Cas Number : 7017-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-110
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-91852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91852
Availability details for
Quantity
  • Availability
  • Add to Cart