ENA341295287 Enamine

1-{[3-(1-ethoxyethyl)-1,2,4-oxadiazol-5-yl]methyl}-1,4-diazepane

  • MDL number MFCD20181700
  • Cas Number : 1354959-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91859
Availability details for
Quantity
  • Availability
  • Add to Cart