ENA341295289 Enamine

5-bromo-2-hydroxybenzohydrazide

  • MDL number MFCD00017567
  • Cas Number : 39635-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-91861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91861
Availability details for
Quantity
  • Availability
  • Add to Cart