ENA341295294 Enamine

N-benzyl-N-methylpyrrolidin-3-amine

  • MDL number MFCD16709668
  • Cas Number : 1220172-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91871

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91871
Availability details for
Quantity
  • Availability
  • Add to Cart