ENA341295296 Enamine

3-(1H-imidazol-1-yl)-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD14632033
  • Cas Number : 1249241-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91877
Availability details for
Quantity
  • Availability
  • Add to Cart