ENA341295299 Enamine

bis(2,2,2-trifluoroethyl) oxalate

  • MDL number MFCD18380784
  • Cas Number : 466684-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90151
Availability details for
Quantity
  • Availability
  • Add to Cart