ENA341295301 Enamine

ethyl 2,2,2-trifluoroethyl oxalate

  • MDL number MFCD18380785
  • Cas Number : 1269151-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90152
Availability details for
Quantity
  • Availability
  • Add to Cart