ENA341295302 Enamine

5-[2-(difluoromethoxy)phenyl]-1,2,4-triazin-3-amine

  • MDL number MFCD11134056
  • Cas Number : 1094454-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91887
Availability details for
Quantity
  • Availability
  • Add to Cart