ENA341295304 Enamine

1-(1-phenylpropyl)piperazine

  • MDL number MFCD10032769
  • Cas Number : 944070-19-3
  • Molecular Weight : 204.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-91891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91891
Availability details for
Quantity
  • Availability
  • Add to Cart