ENA341295305 Enamine

2-methyl-3-sulfanylpropan-1-ol

  • MDL number MFCD01366158
  • Cas Number : 56160-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-91892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91892
Availability details for
Quantity
  • Availability
  • Add to Cart