ENA341295311 Enamine

4-[(hydrazinecarbonyl)amino]benzoic acid

  • MDL number MFCD08700381
  • Cas Number : 73469-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91933
Availability details for
Quantity
  • Availability
  • Add to Cart