ENA341295313 Enamine

cyclobutanecarboxamide

  • MDL number MFCD00029076
  • Cas Number : 1503-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-155
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91936
Availability details for
Quantity
  • Availability
  • Add to Cart