ENA341295316 Enamine

2-bromo-1-[4-(propan-2-yl)phenyl]propan-1-one

  • MDL number MFCD12094680
  • Cas Number : 820260-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91939
Availability details for
Quantity
  • Availability
  • Add to Cart