ENA341295334 Enamine

1-methyl-5-phenyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD12116656
  • Cas Number : 400715-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91968

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91968
Availability details for
Quantity
  • Availability
  • Add to Cart