ENA341295337 Enamine

2-(piperidin-4-yl)aniline

  • MDL number MFCD18483349
  • Cas Number : 255050-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90158
Availability details for
Quantity
  • Availability
  • Add to Cart