ENA341295347 Enamine

4,6-dichloro-2-cyclopropyl-5-methylpyrimidine

  • MDL number MFCD18844042
  • Cas Number : 21721-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92015
Availability details for
Quantity
  • Availability
  • Add to Cart