ENA341295349 Enamine

1-(4,6-dimethylpyrimidin-2-yl)-3-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12633281
  • Cas Number : 1152909-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92017

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92017
Availability details for
Quantity
  • Availability
  • Add to Cart