ENA341295352 Enamine

3-(5-methyl-1,3,4-oxadiazol-2-yl)benzoic acid

  • MDL number MFCD11935905
  • Cas Number : 915707-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92020
Availability details for
Quantity
  • Availability
  • Add to Cart