ENA341295358 Enamine

1-[(4-aminophenyl)methyl]-3-ethylurea

  • MDL number MFCD12763341
  • Cas Number : 1183730-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92027
Availability details for
Quantity
  • Availability
  • Add to Cart