ENA341295373 Enamine

2-methanesulfonyl-N-methylcyclopentan-1-amine

  • MDL number MFCD16871260
  • Cas Number : 1343125-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92046

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92046
Availability details for
Quantity
  • Availability
  • Add to Cart