ENA341295374 Enamine

3-[3-(1-ethoxyethyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD20346349
  • Cas Number : 1340179-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92047

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92047
Availability details for
Quantity
  • Availability
  • Add to Cart