ENA341295399 Enamine

4-(chloromethyl)-1-methoxy-2-[(trifluoromethyl)sulfanyl]benzene

  • MDL number MFCD20441629
  • Cas Number : 1354950-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90171
Availability details for
Quantity
  • Availability
  • Add to Cart