ENA341295405 Enamine

2-amino-6-methyl-5,6,7,8-tetrahydro-1,6-naphthyridine-3-carbonitrile

  • MDL number MFCD20232416
  • Cas Number : 1334490-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92126
Availability details for
Quantity
  • Availability
  • Add to Cart