ENA341295407 Enamine

methyl 5-chloro-2-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

  • MDL number MFCD20501993
  • Cas Number : 1039037-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92128
Availability details for
Quantity
  • Availability
  • Add to Cart