ENA341295420 Enamine

methyl 2-chloro-2-(2,4-dichlorophenyl)acetate

  • MDL number MFCD16694239
  • Cas Number : 225225-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92147
Availability details for
Quantity
  • Availability
  • Add to Cart