ENA341295421 Enamine

[2-(4-methylphenyl)ethyl](2,2,2-trifluoroethyl)amine

  • MDL number MFCD12776022
  • Cas Number : 1182820-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92148
Availability details for
Quantity
  • Availability
  • Add to Cart