ENA341295424 Enamine

N'-(2-chloroacetyl)-2,2-dimethylpropanehydrazide

  • MDL number MFCD18916812
  • Cas Number : 40016-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92191
Availability details for
Quantity
  • Availability
  • Add to Cart