ENA341295427 Enamine

2-[4-(4-fluorophenyl)-1H-imidazol-2-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD14583823
  • Cas Number : 1354954-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90178
Availability details for
Quantity
  • Availability
  • Add to Cart