ENA341295428 Enamine

3-bromo-1-[4-(difluoromethoxy)phenyl]piperidin-2-one

  • MDL number MFCD19590562
  • Cas Number : 1354949-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92195
Availability details for
Quantity
  • Availability
  • Add to Cart