ENA341295429 Enamine

4-[ethyl(phenyl)amino]benzoic acid

  • MDL number MFCD11522113
  • Cas Number : 1039995-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90179
Availability details for
Quantity
  • Availability
  • Add to Cart