ENA341295430 Enamine

ethyl 2-(ethylamino)-1,3-thiazole-4-carboxylate

  • MDL number MFCD20502001
  • Cas Number : 57250-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92198

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92198
Availability details for
Quantity
  • Availability
  • Add to Cart