ENA341295441 Enamine

3-(1,2,3,4-tetrahydroquinolin-1-yl)propan-1-ol

  • MDL number MFCD09971861
  • Cas Number : 88014-20-4
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-92213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92213
Availability details for
Quantity
  • Availability
  • Add to Cart