ENA341295445 Enamine

3-(methanesulfinylmethyl)-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD14613373
  • Cas Number : 1247628-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90185
Availability details for
Quantity
  • Availability
  • Add to Cart