ENA341295446 Enamine

4-bromo-N-(pyrazin-2-yl)benzamide

  • MDL number MFCD12643994
  • Cas Number : 1154240-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-90186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90186
Availability details for
Quantity
  • Availability
  • Add to Cart