ENA341295447 Enamine

4-[bromo(phenyl)methyl]-1,2-difluorobenzene

  • MDL number MFCD12789686
  • Cas Number : 1248278-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92222

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92222
Availability details for
Quantity
  • Availability
  • Add to Cart